accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 123-68-2

IUPAC Name: prop-2-en-1-yl hexanoate
Synonyms: allyl caproate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: RCSBILYQLVXLJG-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results