Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Hexanoate 98.0+%, TCI America™
SDP

Catalog No. A2179500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2179500ML 500 mL
A217925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2179500ML Supplier TCI America Supplier No. A2179500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123-68-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00038339
InChI Key RCSBILYQLVXLJG-UHFFFAOYSA-N
Synonym Hexanoic Acid Allyl Ester
PubChem CID 31266
IUPAC Name prop-2-enyl hexanoate
SMILES CCCCCC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 188°C
Quantity 500 mL
UN Number 2810
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Hexanoate
EINECSNumber (2)-0759
RTECSNumber MO6125000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.