accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-84-2

IUPAC Name: 1-[(2-aminoethyl)amino]propan-2-ol
Synonyms: 1-[(2-aminoethyl)amino]propan-2-ol
Molecular Weight (g/mol): 118.18
Molecular Formula: C5H14N2O
InChi Key: CWKVFRNCODQPDB-UHFFFAOYNA-N
SMILES: CC(O)CNCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results