Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxypropyl)ethylenediamine 96.0+%, TCI America™
SDP

Catalog No. H030025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H030025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H030025G Supplier TCI America Supplier No. H030025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-84-2
Molecular Formula C5H14N2O
Molecular Weight (g/mol) 118.18
MDL Number MFCD00025598
InChI Key CWKVFRNCODQPDB-UHFFFAOYNA-N
Synonym 2-(2-Aminoethylamino)isopropanol, N-Aminoethylisopropanolamine
PubChem CID 101594
IUPAC Name 1-[(2-aminoethyl)amino]propan-2-ol
SMILES CC(O)CNCCN

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
UN Number 2735
Formula Weight 118.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxypropyl)ethylenediamine
EINECSNumber (2)-0337
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.