accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSOHO

CAS RN 123-93-3

IUPAC Name: 2-[(carboxymethyl)sulfanyl]acetic acid
Synonyms: thiodiglycolic acid usaf E-2 thiodiglycollic acid usaf CB-36 dicarboxymethyl sulfide TDGA
Molecular Weight (g/mol): 150.15
Molecular Formula: C4H6O4S
InChi Key: UVZICZIVKIMRNE-UHFFFAOYSA-N
SMILES: OC(=O)CSCC(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results