accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 123-99-9

IUPAC Name: nonanedioic acid
Synonyms: azelaic acid acido azelaico technical grade azelaic acid azelaic acid, technical grade lepargylate anchoate azelex azalaic acid poly(azelaic anhydride) emerox 1110
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: BDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (13)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (8)
Quantity 
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (4)
  • (11)
  • (4)
  • (13)
  • (1)
  • (1)
  • (3)
  • (3)
  • (8)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)