accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123053-23-6

IUPAC Name: 2-amino-3-(4-chlorophenyl)propanoic acid hydrochloride
Synonyms: fenclonine hydrochloride
Molecular Weight (g/mol): 236.09
Molecular Formula: C9H11Cl2NO2
InChi Key: PFOCEDBJFKVRHU-UHFFFAOYNA-N
SMILES: Cl.NC(CC1=CC=C(Cl)C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results