Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-L-phenylalanine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C17095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17095G Supplier TCI America Supplier No. C17095G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123053-23-6
Molecular Formula C9H11Cl2NO2
Molecular Weight (g/mol) 236.092
MDL Number MFCD03095606
InChI Key PFOCEDBJFKVRHU-QRPNPIFTSA-N
Synonym (S)-2-Amino-3-(4-chlorophenyl)propionic Acid Hydrochloride, H-Phe(4-Cl)-OH.HCl
PubChem CID 12647247
IUPAC Name (2S)-2-amino-3-(4-chlorophenyl)propanoic acid;hydrochloride
SMILES C1=CC(=CC=C1CC(C(=O)O)N)Cl.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 236.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-L-phenylalanine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.