accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2NH2CH3SOOOH

CAS RN 1231709-21-9

IUPAC Name: 4-methylbenzene-1-sulfonic acid; ethyl 2-aminopent-4-enoate
Synonyms: ethyl 2-aminopent-4-enoate; para-toluene sulfonate
Molecular Weight (g/mol): 315.38
Molecular Formula: C14H21NO5S
InChi Key: MEQLYDZSCPUCRY-UHFFFAOYNA-N
SMILES: CCOC(=O)C(N)CC=C.CC1=CC=C(C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results