Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-alpha-Allylglycine ethyl ester p-toluenesulfonate, 98%, 98% ee, Thermo Scientific™

Catalog No. AC441280010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441280010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441280010 Supplier Thermo Scientific Chemicals Supplier No. 441280010
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1231709-21-9
Molecular Formula C7H13NO2·C7H8O3S
Molecular Weight (g/mol) 315.39
InChI Key MEQLYDZSCPUCRY-RGMNGODLSA-N
Synonym s-allylglycine ethyl ester p-toluenesulfonateee, ethyl 2s-2-amino-4-pentenoate 4-methylbenzenesulfonate 1:1
PubChem CID 119026235
IUPAC Name ethyl (2S)-2-aminopent-4-enoate;4-methylbenzenesulfonic acid
SMILES CCOC(=O)C(CC=C)N.CC1=CC=C(C=C1)S(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 131°C to 133°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 13
Formula Weight 315.39
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-α-Allylglycine ethyl ester p-toluenesulfonate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.