accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CCH3NOONOH3CCH3CH3H3COOOOO(?)(?)

CAS RN 123174-58-3

IUPAC Name: 7,18-bis[2,6-bis(propan-2-yl)phenyl]-11,14,22,26-tetraphenoxy-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: 7,18-bis(2,6-diisopropylphenyl)-11,14,22,26-tetraphenoxy-7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Weight (g/mol): 1079.26
Molecular Formula: C72H58N2O8
InChi Key: ZZSIDSMUTXFKNS-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1C(=O)C2=CC(OC3=CC=CC=C3)=C3C4=C(OC5=CC=CC=C5)C=C5C(=O)N(C(=O)C6=CC(OC7=CC=CC=C7)=C(C7=C(OC8=CC=CC=C8)C=C(C1=O)C2=C37)C4=C56)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results