Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2,6-diisopropylphenyl)-1,6,7,12-tetraphenoxy-3,4,9,10-perylenetetracarboxylic Diimide 95.0+%, TCI America™
SDP

Catalog No. B43945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43945G 5 g
B43941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43945G Supplier TCI America Supplier No. B43945G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123174-58-3
Molecular Formula C72H58N2O8
Molecular Weight (g/mol) 1079.262
InChI Key ZZSIDSMUTXFKNS-UHFFFAOYSA-N
PubChem CID 16172388
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C(=O)C3=CC(=C4C5=C(C=C6C7=C5C(=C(C=C7C(=O)N(C6=O)C8=C(C=CC=C8C(C)C)C(C)C)OC9=CC=CC=C9)C1=C(C=C(C3=C41)C2=O)OC1=CC=CC=C1)OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Red
Quantity 5 g
Formula Weight 1079.26
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2,6-diisopropylphenyl)-1,6,7,12-tetraphenoxy-3,4,9,10-perylenetetracarboxylic Diimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.