accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123191-00-4

IUPAC Name: 1-[tris(propan-2-yl)silyl]-1H-indole
Synonyms: 1-(triisopropylsilyl)indole
Molecular Weight (g/mol): 273.5
Molecular Formula: C17H27NSi
InChi Key: YQGQSALLXQDVFW-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results