Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Triisopropylsilyl)indole 94.0+%, TCI America™
SDP

Catalog No. T18221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18221G Supplier TCI America Supplier No. T18221G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123191-00-4
Molecular Formula C17H27NSi
Molecular Weight (g/mol) 273.495
MDL Number MFCD02093498
InChI Key YQGQSALLXQDVFW-UHFFFAOYSA-N
PubChem CID 11459999
IUPAC Name indol-1-yl-tri(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=CC=CC=C21

Specifications

Quantity 1 g
Formula Weight 273.50
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Triisopropylsilyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.