accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123291-97-4

IUPAC Name: (2-ethoxy-5-methylphenyl)boronic acid
Synonyms: 2-ethoxy-5-methylphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: KBGGTPGSWHNQEX-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C)C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results