Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1189-5G 5 g
E11891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1189-5G Supplier TCI America Supplier No. E11895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123291-97-4
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD00196832
InChI Key KBGGTPGSWHNQEX-UHFFFAOYSA-N
PubChem CID 4615191
IUPAC Name (2-ethoxy-5-methylphenyl)boronic acid
SMILES CCOC1=C(C=C(C)C=C1)B(O)O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Synonym 2-ethoxy-5-methylbenzeneboronic acid, 2-ethoxy-5-methylphenyl boronic acid, 2-ethoxy-5-methyl-phenyl boronic acid, 2-ethoxy-5-methylphenylboronicacid, 4-ethoxytoluene-3-boronic acid
Formula Weight 180.01
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.