accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNN

CAS RN 123333-53-9

IUPAC Name: 1H-1,2,3-benzotriazol-1-ol
Synonyms: hydroxybenzotriazole|benzotriazole,2-hydroxy
Molecular Weight (g/mol): 135.13
Molecular Formula: C6H5N3O
InChi Key: ASOKPJOREAFHNY-UHFFFAOYSA-N
SMILES: ON1N=NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results