accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123333-85-7

IUPAC Name: lithium(1+) cyanosulfanide
Synonyms: lithium(1+) thiocyanate
Molecular Weight (g/mol): 65.02
Molecular Formula: CLiNS
InChi Key: ZJZXSOKJEJFHCP-UHFFFAOYSA-M
SMILES: [Li+].[S-]C#N
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)