accessibility menu, dialog, popup

UserName

Viewing profile as user:
CrFFF

CAS RN 123333-98-2

IUPAC Name: chromium(3+) trifluoride
Synonyms: chromium trifluoride
Molecular Weight (g/mol): 108.99
Molecular Formula: CrF3
InChi Key: FTBATIJJKIIOTP-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Cr+3]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results