accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOHOPOOHHONH2NH2NH2H2O

CAS RN 123334-13-4

IUPAC Name: 2-(phosphonooxy)prop-2-enoic acid tris(cyclohexanamine) hydrate
Synonyms: tris(cyclohexylamine) phosphoenolpyruvic acid hydrate
Molecular Weight (g/mol): 483.59
Molecular Formula: C21H46N3O7P
InChi Key: CWNPPSJOSPTFGQ-UHFFFAOYSA-N
SMILES: O.NC1CCCCC1.NC1CCCCC1.NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results