Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic acid tris(cyclohexylammonium) salt monohydrate, 97+%, Thermo Scientific™

Catalog No. AAA1731703
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1731703 1 g
AAA1731701 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1731703 Supplier Thermo Scientific Chemicals Supplier No. A1731703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-13-4
Molecular Formula C21H46N3O7P
Molecular Weight (g/mol) 483.587
MDL Number MFCD00004258
InChI Key CWNPPSJOSPTFGQ-UHFFFAOYSA-N
Synonym tri cyclohexylamine salt, 3c6h13n.c3h5o6p.h2o, cyclohexanamine,2-phosphonooxyprop-2-enoic acid,hydrate, phosphoenolpyruvic acid tri cyclohexylamine salt monohydrate, phosphoenolpyruvic acid tris cyclohexylammonium salt hydrate, tris cyclohexylamine phosphoenolpyruvic acid hydrate, 2-phosphonooxy prop-2-enoic acid-cyclohexanamine-water 1/3/1
PubChem CID 71311207
IUPAC Name cyclohexanamine;2-phosphonooxyprop-2-enoic acid;hydrate
SMILES C=C(C(=O)O)OP(=O)(O)O.C1CCC(CC1)N.C1CCC(CC1)N.C1CCC(CC1)N.O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 1 g
Formula Weight 483.59 (465.57 Anhydrous)
Chemical Name or Material Phosphoenolpyruvic acid tris(cyclohexylammonium) salt monohydrate
Recommended Storage −30° to −10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.