accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHOHOHH2O

CAS RN 123334-16-7

IUPAC Name: tetrahydroxycyclohexa-2,5-diene-1,4-dione hydrate
Synonyms: kelox hydrate
Molecular Weight (g/mol): 190.11
Molecular Formula: C6H6O7
InChi Key: CJFTUKFVMMYYHH-UHFFFAOYSA-N
SMILES: O.OC1=C(O)C(=O)C(O)=C(O)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)