Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydroxy-1,4-benzoquinone Hydrate 96.0+%, TCI America™
SDP

Catalog No. T10905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10905G Supplier TCI America Supplier No. T10905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123334-16-7
Molecular Formula C6H6O7
Molecular Weight (g/mol) 190.107
MDL Number MFCD00001597
InChI Key CJFTUKFVMMYYHH-UHFFFAOYSA-N
Synonym tetrahydroxy-1,4-benzoquinone hydrate, 2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione hydrate, tetrahydroxy-1,4-quinone hydrate, tetrahydroxy-1,4-quinone monohydrate, kelox hydrate, 2,5-cyclohexadiene-1,4-dione, 2,3,5,6-tetrahydroxy-, hydrate, thq hydrate, prestwick_964, acmc-209aoq, tetrahydroxy-1,4-benzoquinonehydrate
PubChem CID 6420048
IUPAC Name 2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione;hydrate
SMILES C1(=C(C(=O)C(=C(C1=O)O)O)O)O.O

Specifications

Quantity 5 g
Formula Weight 172.09
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydroxy-1,4-benzoquinone Hydrate
EINECSNumber (9)-2072
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.