accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12354-85-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 618.07
Molecular Formula: C20H30Cl4Rh2
InChi Key: QNIVKTTWBMFSBR-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Rh++].[Rh++].C[C]1C(C)=C(C)C(C)=C1C.C[C]1C(C)=C(C)C(C)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)