Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentamethylcyclopentadienylrhodium(III) Chloride Dimer

Catalog No. 338370250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-837-0250MG 250 mg
33-837-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 33-837-0250MG Supplier Sigma Aldrich Supplier No. 338370250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Umicore Precatalysts for Asymmetric and Cross-Coupling CatalysisCatalyst for:Oxidative olefination reactionsC-C bond cleavage of secondary alcoholsOrtho C-H olefination of phenol derivativesOxidative annulation of pyridinesOxidative ortho-acylation of benzamides with aldehydes via direct functionalization of the sp2 C-H bondAmidation reactionsReductive alkylation reactions

Specifications

CAS 12354-85-7
Molecular Formula C20H30Cl4Rh2
Linear Formula [Rh(C5Me5)Cl2]2
MDL Number MFCD00061552
Quantity 250 mg
Synonym 1,3-Cyclopentadiene, 1,2,3,4,5-pentamethyl-, rhodium complex; Bis(dichloro(pentamethylcyclopentadienyl)rhodium); Bis(pentamethylcyclopentadienylrhodium dichloride); Dichloro(pentamethylcyclopentadienyl)rhodium(II) dimer; Tetrachlorobis(pentamethylcyclopentadienyl)dirhodium
Molecular Weight (g/mol) 618.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.