accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFFNH2

CAS RN 123572-58-7

IUPAC Name: 2-fluoro-4-(trifluoromethoxy)aniline
Synonyms: 2-fluoro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: HDDJDTDRGKHRBG-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results