accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNFOH

CAS RN 123843-57-2

IUPAC Name: 2,6-difluoro-4-hydroxybenzonitrile
Synonyms: 2,6-difluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: KEIYYIGMDPTAPL-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(C#N)C(F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results