Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-4-hydroxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D34245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34245G Supplier TCI America Supplier No. D34245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123843-57-2
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD03094499
InChI Key KEIYYIGMDPTAPL-UHFFFAOYSA-N
Synonym 4-Cyano-3,5-difluorophenol
PubChem CID 2778774
IUPAC Name 2,6-difluoro-4-hydroxybenzonitrile
SMILES C1=C(C=C(C(=C1F)C#N)F)O

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 155.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.