accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PCH3H3CAgI

CAS RN 12389-34-3

IUPAC Name: iodosilver; trimethylphosphane
Synonyms: silver iodide; trimethylphosphine
Molecular Weight (g/mol): 310.85
Molecular Formula: C3H9AgIP
InChi Key: UEZMPJVLYIQMMG-UHFFFAOYSA-M
SMILES: [Ag]I.CP(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results