Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylphosphine-silver Iodide Complex, Thermo Scientific™

Catalog No. AC213460250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC213460250 25 g
AC213460050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC213460250 Supplier Thermo Scientific Chemicals Supplier No. 213460250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12389-34-3
Molecular Formula C3H9AgIP
Molecular Weight (g/mol) 310.851
InChI Key UEZMPJVLYIQMMG-UHFFFAOYSA-M
Synonym iodosilver; trimethylphosphane, trimethylphosphine silver iodide complex, silver iodide; trimethylphosphine
PubChem CID 357596
IUPAC Name iodosilver;trimethylphosphane
SMILES CP(C)C.[Ag]I

Specifications

Linear Formula [(CH3)3P.AgI]4
Quantity 25 g
Formula Weight 1243.41
Chemical Name or Material Trimethylphosphine-silver iodide complex
Hazard Category STENCH
Hazard Statement LIGHT SENSITIVE
Precautionary Statement HEAT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.