CAS RN 1239-45-8
Ethidium Bromide Solution, MP Biomedicals
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Ethidium bromide, 95%
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)
Ethidium Bromide Solution (10mg/mL), Molecular biology grade
Ethidium bromide soln., 10mg/ml
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium bromide, 95%, pure
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium bromide, >98%, Molecular Biology Reagent, MP Biomedicals™
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Thermo Scientific Chemicals Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Synonym: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Thermo Scientific Chemicals Ethidium Bromide, Molecular Biology Grade, 98%
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Synonym: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; Homidium bromide
Ethidium bromide soln., 0.625mg/ml
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium Bromide 95.0+%, TCI America™
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium Bromide, Fisher BioReagents
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Ethidium Bromide, DNase, RNase and Protease Free 99+%, Thermo Scientific™
CAS: 1239-45-8 Molecular Formula: C21H20BrN3 Molecular Weight (g/mol): 394.32 MDL Number: MFCD00011724 InChI Key: ZMMJGEGLRURXTF-UHFFFAOYSA-N Synonym: ethidium bromide,homidium bromide,dromilac,3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide,ethydium bromide,3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide,etbr,2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide,2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide,unii-059nuo2z1l PubChem CID: 14710 ChEBI: CHEBI:4883 IUPAC Name: 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide SMILES: [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12