Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethidium Bromide, Fisher BioReagents

Catalog No. BP1025 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP102-5 5 g Amber Glass
BP102-1 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP102-5

missing translation for 'validMainframeMsg'

  • Ethidium bromide (EtBr)  commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light
  • Ethidium bromide intercalates double-stranded DNA and RNA
  • Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer 
  • EtBr also acts as inhibitor of RNA and DNA polymerase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 260°C
Color Purple
pH 4.0 to 7.0
Loss on Drying 5% max.
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 4769
Formula Weight 394.32
Physical Form Powder or Solid
Chemical Name or Material Ethidium Bromide
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.