accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCrONNOSOOOONNOSOOO(E/Z)(E/Z)

CAS RN 12392-64-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 880.73
Molecular Formula: C40H22CrN4Na2O10S2
InChi Key: LCZPIYCNOWJWPQ-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Cr+3].[O-]C1=CC=C2C=CC=CC2=C1N=NC1=C2C=CC=CC2=C(C=C1[O-])S([O-])(=O)=O.[O-]C1=CC=C2C=CC=CC2=C1N=NC1=C2C=CC=CC2=C(C=C1[O-])S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results