Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Blue 161, MP Biomedicals™

Catalog No. ICN15259925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15259925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15259925 Supplier MP Biomedicals, Inc Supplier No. 0215259925

Soluble in water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12392-64-2
Molecular Formula C40H27CrN4Na2O10S2+
Molecular Weight (g/mol) 885.77
MDL Number MFCD00151053
InChI Key SUXIOICJRRGPND-UHFFFAOYSA-M
Synonym Disazo, Palatine Fast Blue BN
PubChem CID 73555498
IUPAC Name disodium;chromium;hydron;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES [H+].C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+].[Na+].[Cr]

Specifications

Formula Weight 416.39 + xCr
Quantity 25 g
Chemical Name or Material Acid Blue 161
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.