accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiOOOCH3OCH3

CAS RN 1239602-38-0

IUPAC Name: trimethoxy({8-[(oxiran-2-yl)methoxy]octyl})silane
Synonyms: trimethoxy[8-(oxiran-2-ylmethoxy)octyl]silane
Molecular Weight (g/mol): 306.47
Molecular Formula: C14H30O5Si
InChi Key: QYJYJTDXBIYRHH-UHFFFAOYNA-N
SMILES: CO[Si](CCCCCCCCOCC1CO1)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results