Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[8-(Glycidyloxy)-n-octyl]trimethoxysilane 97.0+%, TCI America™
SDP

Catalog No. G04695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04695G 5 g
G046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04695G Supplier TCI America Supplier No. G04695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1239602-38-0
Molecular Formula C14H30O5Si
Molecular Weight (g/mol) 306.474
InChI Key QYJYJTDXBIYRHH-UHFFFAOYSA-N
PubChem CID 102529357
IUPAC Name trimethoxy-[8-(oxiran-2-ylmethoxy)octyl]silane
SMILES CO[Si](CCCCCCCCOCC1CO1)(OC)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 306.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material [8-(Glycidyloxy)-n-octyl]trimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.