accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-11-8

IUPAC Name: non-1-ene
Synonyms: nonene isononene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: JRZJOMJEPLMPRA-UHFFFAOYSA-N
SMILES: CCCCCCCC=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results