Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonene 99.5+%, TCI America™
SDP

Catalog No. S03395ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03395ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03395ML Supplier TCI America Supplier No. S03395ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124-11-8
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00009568
InChI Key JRZJOMJEPLMPRA-UHFFFAOYSA-N
PubChem CID 31285
ChEBI CHEBI:77443
IUPAC Name non-1-ene
SMILES CCCCCCCC=C

Specifications

Physical Form Liquid
UN Number 3295
Color Colorless
Quantity 5 mL
Formula Weight 126.24
Percent Purity ≥99.5% (GC)
Chemical Name or Material 1-Nonene [Standard Material for GC]
EINECSNumber (2)-0025
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.