accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-12-9

IUPAC Name: octanenitrile
Synonyms: octanonitrile
Molecular Weight (g/mol): 125.22
Molecular Formula: C8H15N
InChi Key: YSIMAPNUZAVQER-UHFFFAOYSA-N
SMILES: CCCCCCCC#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)