Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptyl Cyanide 97%, Thermo Scientific™

Catalog No. AC156261000
Change view
Click to view available options
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC156261000 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156261000 Supplier Thermo Scientific Chemicals Supplier No. 156261000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 124-12-9
CAS Min % 96.0
CAS Max % 100.0
Density 0.81g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H15N
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Linear Formula CH3(CH2)6CN
Quantity 100mL
MDL Number MFCD00001983
Beilstein 02, 349
Synonym caprylonitrile, octanonitrile, arneel 8, 1-cyanoheptane, caprylnitrile, n-heptyl cyanide, heptyl cyanide, heptylcyanide, unii-d6646v87py, n-heptylcyanide
InChI Key YSIMAPNUZAVQER-UHFFFAOYSA-N
SMILES CCCCCCCC#N
IUPAC Name octanenitrile
Molecular Weight (g/mol) 125.22
PubChem CID 31286
Formula Weight 125.21
Percent Purity ≥96.0%
Chemical Name or Material n-Heptyl cyanide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-682-2
RTECSNumber RG9700300
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.