accessibility menu, dialog, popup

UserName

Viewing profile as user:
OCO

CAS RN 124-38-9

IUPAC Name: methanedione
Synonyms: carbon dioxide 二氧化碳 carbonic anhydride
Molecular Weight (g/mol): 44.01
Molecular Formula: CO2
InChi Key: CURLTUGMZLYLDI-UHFFFAOYSA-N
SMILES: O=C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results