accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)OH(R)F(S)O(S)CH3(S)(R)HO(S)HOOHO

CAS RN 124-94-7

IUPAC Name: (1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,2,10-trihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: triamcinolone nasacort kenalog nasacort aq aristocort fougera azmacort vetalog triderm kenacort
Molecular Weight (g/mol): 394.44
Molecular Formula: C21H27FO6
InChi Key: GFNANZIMVAIWHM-OBYCQNJPSA-N
SMILES: C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results