accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFFBr

CAS RN 1240257-47-9

IUPAC Name: 1-(bromomethyl)-2-fluoro-4-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-2-fluoro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H5BrF4O
InChi Key: NLXMTHLJHXYXJN-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results