Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Catalog No. AAH34446MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34446MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH34446MD Supplier Thermo Scientific Chemicals Supplier No. H34446MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1240257-47-9
Molecular Formula C8H5BrF4O
Quantity 250 mg
MDL Number MFCD16652442
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 2-fluoro-4-trifluoromethoxy benzyl bromide, 1-bromomethyl-2-fluoro-4-trifluoromethoxy benzene
InChI Key NLXMTHLJHXYXJN-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)CBr
IUPAC Name 1-(bromomethyl)-2-fluoro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol) 273.025
PubChem CID 66523446
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzyl bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.