accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONSNNCH3

CAS RN 1240328-26-0

IUPAC Name: ethyl 4-methyl-2-(1H-pyrazol-1-yl)-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-2-(pyrazol-1-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 237.28
Molecular Formula: C10H11N3O2S
InChi Key: QIEQTJFJYQVLIU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results