accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNHOOHHO(E/Z)(E/Z)

CAS RN 124061-43-4

IUPAC Name: 4-hydroxy-6-{[(2-{[(3-hydroxy-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}ethyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Synonyms: 4-hydroxy-6-{[(2-{[(3-hydroxy-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}ethyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 300.31
Molecular Formula: C16H16N2O4
InChi Key: KNARIIFAOCMXAR-UHFFFAOYSA-N
SMILES: OC1=CC(=CNCCNC=C2C=C(O)C=CC2=O)C(=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results