Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(5-hydroxysalicylidene)ethylenediamine 98.0+%, TCI America™
SDP

Catalog No. B30155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30155G 5 g
B30151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30155G Supplier TCI America Supplier No. B30155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 124061-43-4
Molecular Formula C16H16N2O4
Molecular Weight (g/mol) 300.314
MDL Number MFCD04039747
InChI Key KNARIIFAOCMXAR-HWAYABPNSA-N
Synonym n,n'-bis 5-hydroxysalicylidene ethylenediamine, n,n'-ethylenebis 5-hydroxysalicylideneimine, n,n-bis 5-hydroxysalicylidene ethylenediamine, 2,2'-ethane-1,2-diylbis azanylylidene bis methanylylidene bis benzene-1,4-diol
PubChem CID 24985320
IUPAC Name (6Z)-4-hydroxy-6-[[2-[[(Z)-(3-hydroxy-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=CNCCNC=C2C=C(C=CC2=O)O)C=C1O

Specifications

Melting Point 250°C
Quantity 5 g
Formula Weight 300.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Powder
Chemical Name or Material N,N′-Bis(5-hydroxysalicylidene)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.