accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124236-29-9

IUPAC Name: N-ethyl-N-(2-methylphenyl)but-2-enamide
Synonyms: eurax eurasil epa pesticide chemical code 055701 euraxil eurax lotion caswell no. 431B veteusan crotamitex eurax cream crotamiton (product)
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: DNTGGZPQPQTDQF-UHFFFAOYSA-N
SMILES: CCN(C(=O)C=CC)C1=CC=CC=C1C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)