Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Crotamiton, 97%

Catalog No. AAH5918306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5918306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5918306 Supplier Thermo Scientific Chemicals Supplier No. H5918306

missing translation for 'validMainframeMsg'

CAS: 124236-29-9 | C13H17NO | 203.29 g/mol

Crotamiton was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2. N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crotamiton was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2. N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.

Solubility
Soluble in water (1:500), alcohol, methanol, ether, and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124236-29-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.29
MDL Number MFCD00026989
InChI Key DNTGGZPQPQTDQF-XBXARRHUSA-N
Synonym crotamiton, n-ethyl-o-crotonotoluidide, crotamitone, eurax, veteusan, eurasil, euraxil, trans-crotamiton, crotamitonum, crotan
PubChem CID 688020
IUPAC Name (E)-N-ethyl-N-(2-methylphenyl)but-2-enamide
SMILES CCN(C(=O)\C=C\C)C1=CC=CC=C1C

Specifications

Density 0.987
Boiling Point 153°C to 155°C (13 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.5405
Linear Formula CH3CH=CHCON(C2H5)C6H4CH3
Quantity 5 g
Solubility Information Soluble in water (1:500),alcohol,methanol,ether,and ethanol.
Formula Weight 203.28
Percent Purity 97%
Chemical Name or Material Crotamiton
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
RTECSNumber GQ7000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.