accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH4FPFFFFFCoCHCH

CAS RN 12427-42-8

IUPAC Name: bis(cyclopenta-2,4-dien-1-uid-1-yl)cobalttris(ylium); hexafluoro-λ⁵-phosphanuide; methane
Synonyms: bis(cyclopenta-2,4-dien-1-uid-1-yl)cobalttris(ylium) methane hexafluorophosphate
Molecular Weight (g/mol): 350.13
Molecular Formula: C11H14CoF6P
InChi Key: UQKPUAVRBNGAFO-UHFFFAOYSA-N
SMILES: C.F[P-](F)(F)(F)(F)F.[Co+3]([CH-]1C=CC=C1)[CH-]1C=CC=C1
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)